Lithium peroxide ≥95%

Supplier: Thermo Scientific Chemicals
Danger

AC38988-0100
TS38988-0100EA 53.43 USD
TS38988-0100
Lithium peroxide ≥95%
Lithium peroxide
Formula: Li₂O₂
MW: 45.88 g/mol
Density: 1.2063 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016184
CAS Number: 12031-80-0
UN: 1472
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White to light yellow Powder
Titration with KMnO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR