Potassium nitrite 96+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC42306-0025
AC42306-0050
AC42306-5000
TS42306-0025EA
383
USD
TS42306-0025
TS42306-5000
TS42306-0050
Potassium nitrite 96+% ACS
Potassium nitrite
Formula:
KNO₂ MW: 85.1 g/mol Melting Pt: 387 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011408 CAS Number: 7758-09-0 EINECS: 231-832-4 UN: 1488 ADR: 5.1,II Merck Index: 12,07816 |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Titration with KMnO4 | ≥96.0 % |
Chloride (Cl) | ≤300 ppm |
Sulfate (SO4) | ≤100 ppm |
Calcium (Ca) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Magnesium (Mg) | ≤20 ppm |
Sodium (Na) | ≤5000 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...