Potassium bromate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC20885-0025 AC20885-5000
TS20885-0025EA 517 USD
TS20885-0025 TS20885-5000
Potassium bromate 99+%, Extra Pure
Potassium bromate
Formula: KBrO₃
MW: 167 g/mol
Melting Pt: 350 °C
Density: 3.27 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Titration Iodometric ≥99.0 %
Bromide (Br) ≤500 ppm
Sulfate (SO4) ≤100 ppm
Lead (Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR