Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane 95+%

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethylbis(2,3,4,5-tetramethyl-2,4-cyclopentadien-1-yl) silanedimethyl-bis(2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl)silaneSilane, dimethylbis(tetramethylcyclopentadienyl)- (mixed isomers)

AC38890-5000
TS38890-5000EA 203.63 USD
TS38890-5000
Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane 95+%
Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
Formula: C₂₀H₃₂Si
Melting Pt: 65 - 68 °C
MDL Number: MFCD00674017
CAS Number: 89597-05-7

Order Now

Specification Test Results

Assay 95% min. (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR