Barium chloride, anhydrous
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC61228-1000
TS61228-1000EA
195.97
USD
TS61228-1000
Barium chloride, anhydrous
Barium chloride
Formula:
BaCl₂ MW: 208.24 g/mol Boiling Pt: 1560 °C (1013 hPa) Melting Pt: ∼963 °C Density: 2.33 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003445 CAS Number: 10361-37-2 UN: 1564 ADR: 6.1,III |
Specification Test Results
Appearance | White Granules |
Assay | ≥97 % (BaCl2) |
Identification | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...