Nickel carbonate

Supplier: Thermo Scientific Chemicals

Synonyms: Nickelous CarbonateNickel(II) carbonate

AC61148-5000 AC61148-1000
TS61148-1000EA 56 USD
TS61148-1000 TS61148-5000
Nickel carbonate
Nickel carbonate
Formula: CNiO₃
MW: 118.7 g/mol
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011144
CAS Number: 3333-67-3

Order Now

Specification Test Results

Appearance Light green Powder
Identification passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR