Cell Meter™ Mitochondrial Autophagy Imaging Kit *Red Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22998
76485-158EA 348.42 USD
76485-158
Cell Meter™ Mitochondrial Autophagy Imaging Kit *Red Fluorescence*, AAT Bioquest
Assays Cellular Assays
Mitochondrial autophagy (also called 'mitophagy') appears to be involved in Alzheimer and Parkinson diseases, mainly induced by the accumulation of depolarized mitochondria.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.