Cell Navigator® Lysosome Staining Kit *Orange Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22657
76483-668EA 173.46 USD
76483-668
Cell Navigator® Lysosome Staining Kit *Orange Fluorescence*, AAT Bioquest
Assays Cellular Assays
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.