1-Aminohydantoin (AHD)-HRP Conjugate (1 mg)
Supplier: AAT Bioquest
50012
76484-038EA 580.89 USD
76484-038
1-Aminohydantoin (AHD)-HRP Conjugate (1 mg)
Proteins and Peptides
1-Aminohydantoin (AHD)-HRP conjugate is used for developing an ELISA assay for quantifying 1-Aminohydantoin (AHD).
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.