iFluor 750™ RGD Conjugate (1 mg)
Supplier: AAT Bioquest
36800
76484-002EA 348.42 USD
76484-002
iFluor 750™ RGD Conjugate (1 mg)
Proteins and Peptides
Near-infrared fluorescence optical imaging (also called in vivo Imaging) is emerging as a powerful technology for studying diseases at molecular level in small animal models.
  • Nir emission (757/779 nm)
  • Brighter and more stable signal
  • Multiple research applications
  • Suitable for both in vivo and in vitro
Caution: For research use only.