Spectinomycin-KLH Conjugate (1 mg)
Supplier: AAT Bioquest
50039
76484-092EA 580.89 USD
76484-092
Spectinomycin-KLH Conjugate (1 mg)
Proteins and Peptides
Spectinomycin-KLH conjugate is used for developing anti-spectinomycin antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.