Sulfathiazol-KLH Conjugate (1 mg)
Supplier: AAT Bioquest
50030
76484-074EA 580.89 USD
76484-074
Sulfathiazol-KLH Conjugate (1 mg)
Proteins and Peptides
Sulfathiazol-KLH conjugate is used for developing anti-sulfathiazol antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.