Sulfadimidine-BSA Conjugate (1 mg)
Supplier: AAT Bioquest
50026
76484-066EA 580.89 USD
76484-066
Sulfadimidine-BSA Conjugate (1 mg)
Proteins and Peptides
Sulfadimidine-BSA conjugate is used for developing anti-sulfadimidine antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.