Cell Meter™ Intracellular GSH Assay Kit *Optimized for Flow Cytometry with 405 nm excitation* (100 Tests), AAT Bioquest
Supplier: AAT Bioquest
22809
76484-860EA 230.87 USD
76484-860
Cell Meter™ Intracellular GSH Assay Kit *Optimized for Flow Cytometry with 405 nm excitation* (100 Tests), AAT Bioquest
Assays Cellular Assays
There are a variety of parameters that can be used for monitoring cell apoptosis.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.