Cell Navigator® Mitochondrion Staining Kit *Orange Fluorescence with 405 nm Excitation* (500 Assays), AAT Bioquest
Supplier: AAT Bioquest
22673
76483-692EA 230.87 USD
76483-692
Cell Navigator® Mitochondrion Staining Kit *Orange Fluorescence with 405 nm Excitation* (500 Assays), AAT Bioquest
Assays Cellular Assays
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labeling sub-cellular organelles such as membranes, lysosomes, mitochondria, nuclei, etc.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.