Cell Navigator® Mitochondrion Staining Kit *Blue Fluorescence* (500 Assays), AAT Bioquest
Supplier: AAT Bioquest
22665
76483-682EA 230.87 USD
76483-682
Cell Navigator® Mitochondrion Staining Kit *Blue Fluorescence* (500 Assays), AAT Bioquest
Assays Cellular Assays
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labeling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.