Spexyte™ Intracellular pH Calibration Buffer Kit (100 Tests), AAT Bioquest
Supplier: AAT Bioquest
21235
76484-784EA 230.87 USD
76484-784
Spexyte™ Intracellular pH Calibration Buffer Kit (100 Tests), AAT Bioquest
Assays Cellular Assays
Intracellular pH (pHi) plays an important modulating role in many cellular events, including cell volume regulation, cellular metabolism, calcium regulation, receptor-mediated signal transduction, ion transport, endocytosis, and other cellular processes.
  • Suitable for all major pH indicators
  • Sensitive and quick results
  • Includes a range of calibration buffers
Caution: For research use only.