Cl-NERF (1 mg), AAT Bioquest
Supplier: AAT Bioquest
21220
76483-362EA 230.87 USD
76483-362
Cl-NERF (1 mg), AAT Bioquest
Stains Microbiology Stains
Cl-NERF is a rhodol dye with pKa = 3.8.
  • pH-dependent fluorescence
  • Ratiometric detection
  • Cell impermeable
Caution: For research use only.