Adapter for FastPrep-96 instruments.
- 2×250 ml tube holder
- For room temperature grinding on FastPrep-96 instruments
Super-size adapter enables FastPrep-96 instruments to accommodate 2×250 ml Lysing Matrix bottles. The ergonomic design of the adapter ensures easy loading of tubes that remain securely sealed during homogenization. A wide range of disposable 250 ml Lysing Matrix bottles ensure thorough homogenization of any sample type in seconds.
The FastPrep bead beating system maximizes mechanical tissue homogenization for efficient isolation of high-quality DNA, RNA and proteins, without the need for chemicals or enzymes.
Caution: Unless specified otherwise, MP Biomedicals' products are for research or further manufacturing use only, not for direct human use.