trFluor™ Anti-IgG Goat Polyclonal Antibody [clone:IgG] trFluor™ Eu, AAT Bioquest
Supplier: AAT Bioquest
16755 16518
76482-470EA 348.42 USD
76482-470 76482-472
trFluor™ Anti-IgG Goat Polyclonal Antibody [clone:IgG] trFluor™ Eu, AAT Bioquest
Antibodies
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • Large stokes shift
  • Improved stability
  • High emission yield
  • Long emission half-life
Caution: For research use only.