ReadiUse™ NADPH Regenerating Kit, AAT Bioquest
Supplier: AAT Bioquest
15265
76482-356EA 348.42 USD
76482-356
ReadiUse™ NADPH Regenerating Kit, AAT Bioquest
Assays Enzyme Assays
NADPH provides the reducing equivalents for biosynthetic reactions and for oxidation-reduction involved in protection against the toxicity of ROS (reactive oxygen species).
  • Ready to use
  • Robust protocol
  • Contains all necessary components
Caution: For research use only.