FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit), AAT Bioquest
Supplier: AAT Bioquest
20001
76482-960EA 114.94 USD
76482-960
FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit), AAT Bioquest
Stains Microscopy and staining reagents
When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.
  • Preserves fluorescence
  • Prevents photobleaching
  • Ready to use
  • Contains all necessary components
Caution: For research use only.