Cal-520N™ potassium salt

Supplier: AAT BIOQUEST INC
Warning

21147
76484-248EA 405.91 USD
76484-248
Cal-520N™ potassium salt
Cal-520N™ potassium salt

Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.


  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell impermeant


Caution: For research use only

MW: 1004,17 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR