Rhod-4™ sodium salt

Supplier: AAT BIOQUEST INC
Warning

21128 21118
76483-270EA 348.42 USD
76483-270 76483-268
Rhod-4™ sodium salt
Rhod-4™ sodium salt

Calcium measurement is critical for numerous biological investigations.


  • Well-cited
  • Long-wavelength emission
  • Cell impermeant


Caution: For research use only

MW: 815,64 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR