Rhod-2 trisodium salt

Supplier: AAT BIOQUEST INC
Warning

21068
76483-212EA 230.87 USD
76483-212
Rhod-2 trisodium salt
Rhod-2 trisodium salt

Calcium measurement is critical for numerous biological investigations.


  • Well-cited
  • Long-wavelength emission
  • Cell impermeant


Caution: For research use only

Formula: C₄₀H₃₉N₄Na₃O₁₁
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR