Fura-8FF™ AM

Supplier: AAT BIOQUEST INC
Warning

20620
76483-104EA 230.87 USD
76483-104
Fura-8FF™ AM
Fura-8FF™ AM

The cell-permeant Fura-8FF AM is an analog of Fura-8 AM with much lower calcium binding affinity, Kd ~10 µm.


  • Calcium-dependent fluorescence
  • Steady signal
  • Cell permeant


Caution: For research use only

MW: 973,86 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR