Cal-590™ AM
Supplier: AAT BIOQUEST INC
|
Warning
|
20511
20512
20510
76483-054EA
348.42
USD
76483-054
76483-052
76483-056
Cal-590™ AM
Cal-590™ AM
Calcium measurement is critical for numerous biological investigations.
- Improved sensitivity
- Improved calcium response
- Long-wavelength emission
Caution: For research use only
MW:
1266,81 g/mol |
ADR:
5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...