7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP

Supplier: AAT BIOQUEST INC
Warning

17092
76485-656EA 387.16 USD
76485-656
7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP
7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP

7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).


  • Ngs building block
  • Useful for multiple applications
  • High quality


Caution: For research use only

Formula: C15H21N8O13P3
MW: 614,3 g/mol
CAS Number: 666847-81-0
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR