7-Deaza-7-Propargylamino-3'-azidomethyl-dATP

Supplier: AAT BIOQUEST INC
Warning

17090
76485-652EA 441.5 USD
76485-652
7-Deaza-7-Propargylamino-3'-azidomethyl-dATP
7-Deaza-7-Propargylamino-3'-azidomethyl-dATP

7-Deaza-7-Propargylamino-3'-azidomethyl-dATP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).


  • Ngs building block
  • Useful for multiple applications
  • High quality


Caution: For research use only

Formula: C15H21N8O12P3
MW: 598,3 g/mol
CAS Number:  666847-93-4
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR