Fluorescein-12-dUTP 1 mM in TRIS buffer (pH 7.5) (Perkin-Elmer)

Supplier: AAT BIOQUEST INC
Warning

17027
76485-482EA 219.43 USD
76485-482
Fluorescein-12-dUTP 1 mM in TRIS buffer (pH 7.5) (Perkin-Elmer)
Fluorescein-12-dUTP

The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.


  • Useful for DNA probe development
  • Well-cited
  • Multiple applications


Caution: For research use only

MW: ~991 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR