Biotin-20-dUTP 1 mM in TRIS buffer (pH 7.5)

Supplier: AAT BIOQUEST INC
Warning

17018
76482-814EA 114.94 USD
76482-814
Biotin-20-dUTP 1 mM in TRIS buffer (pH 7.5)
Biotin-20-dUTP

The biotin-modified deoxyuridine 5'-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3'-end labeling.


  • Useful for DNA probe development
  • Well-cited
  • Multiple applications


Caution: For research use only

MW: 1062,75 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR