TF3-dUTP 1 mM in TRIS buffer (pH 7.5)

Supplier: AAT BIOQUEST INC
Warning

17008
76482-802EA 348.42 USD
76482-802
TF3-dUTP 1 mM in TRIS buffer (pH 7.5)
TF3-dUTP

The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.


  • Useful for fish probe development
  • Well-cited
  • Multiple applications


Caution: For research use only

MW: 1150 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR