Cynomolgus Recombinant CD3E & CD3G Heterodimer Protein (from HEK293 Cells)
Supplier: Sino Biological
CT091-C02H-100 CT091-C02H-1 CT091-C02H-100
76631-462EA 2268 USD
CA76487-772 76631-462 103918-640
Cynomolgus Recombinant CD3E & CD3G Heterodimer Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus CD3E (Q95LI5.2) (Met1-Asp117) was fused with the Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the cynomolgus CD3G (NP_001270839.1) (Met1-Thr113) was fused with the Fc region of human IgG1 at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the cynomolgus CD3E/CD3G heterodimer was purified.