Mouse Recombinant CD79 (CD79A & CD79B Heterodimer) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
CT046-M08H-100
103585-712EA 351.48 USD
103585-712
Mouse Recombinant CD79 (CD79A & CD79B Heterodimer) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CD79A (NP_031681.2) (Met1-Arg137) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding mouse CD79B (NP_031681.2) (Met1-Asp158) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the mouse CD79A and CD79B heterodimer was purified.