Cynomolgus Recombinant B7-H6/B7H6 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90802-C08H-50 90802-C08H-500
76631-638EA 1288 USD
76631-638 103620-522
Cynomolgus Recombinant B7-H6/B7H6 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus NCR3LG1 (XP_005578557.1) (Met1-Asp259) was expressed with a polyhistidine tag at the C-terminus.