Cynomolgus Recombinant FGFR3 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90313-C02H-100
103585-644EA 300.98 USD
103585-644
Cynomolgus Recombinant FGFR3 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus FGFR3 (XP_001101108.1) (Met1-Gly375) was expressed with the Fc region of human IgG1 at the C-terminus.