Cynomolgus Recombinant IL20RB Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90308-K02H-50 90308-K02H-50
103670-620EA 179.78 USD
CA76453-468 103670-620
Cynomolgus Recombinant IL20RB Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus IL20RB (XP_005545913.1) (Asp30-Ala230) was expressed with the Fc region of human IgG1 at the C-terminus.