Cynomolgus Recombinant IL1R1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90279-C08H-100 90279-C08H-1
76632-498EA 1938 USD
76632-498 103585-564
Cynomolgus Recombinant IL1R1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus IL1R1 (EHH61701.1) (Met1-Thr332) was expressed with a polyhistidine tag at the C-terminus.