Cynomolgus Recombinant CD52 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90187-C02H-100 90187-C02H-100
103887-508EA 351.48 USD
CA76477-908 103887-508
Cynomolgus Recombinant CD52 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus CD52 (NP_001253116.1) (Met1-Ser35) was expressed with the Fc region of human IgG1 at the C-terminus.