Cynomolgus Recombinant IL18R1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90121-C08H-1 90121-C08H-100
76631-538EA 1938 USD
76631-538 103585-394
Cynomolgus Recombinant IL18R1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus IL18R1 (F7H8F9) (Met1-Lys315) was expressed with a polyhistidine tag at the C-terminus.