Cynomolgus Recombinant XEDAR/EDA2R Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90110-C02H-100
103585-332EA 300.98 USD
103585-332
Cynomolgus Recombinant XEDAR/EDA2R Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus EDA2R (NP_001180396.1) (Met1-Glu136) was expressed with the Fc region of human IgG1 at the C-terminus.