Rhesus Recombinant EphB1/EPHT2 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90040-C02H-200
103585-178EA 452.48 USD
103585-178
Rhesus Recombinant EphB1/EPHT2 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus EPHB1 (XP_001115263.1) (Met1-Pro540) was expressed with the Fc region of human IgG1 at the C-terminus.