Cynomolgus Recombinant Angiopoietin 1/ANG1/ANGPT1 (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90025-C07H-100 90025-C07H-1
103585-090EA 351.48 USD
103585-090 76632-342
Cynomolgus Recombinant Angiopoietin 1/ANG1/ANGPT1 (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus ANGPT1 (XP_005563980.1) (Arg277-Phe498) was expressed with a polyhistidine tag at the N-terminus.