Rhesus Recombinant CD16/FCGR3 Protein (His & AVI Tag), Biotinylated (from HEK293 Cells)
Supplier: Sino Biological
90013-C27H-B-20
103633-628EA 300.98 USD
103633-628
Rhesus Recombinant CD16/FCGR3 Protein (His & AVI Tag), Biotinylated (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus CD16 (NP_001258584.1) extracellular domain (Met 1- Gln 208) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin–Protein ligase (BirA enzyme) which is co-expressed.