Rat Recombinant B7-H5/GI24/VISTA Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
81347-R02H-200
103585-050EA 452.48 USD
103585-050
Rat Recombinant B7-H5/GI24/VISTA Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat B7H5 (NP_001037765.1) (Met1-Ala191) was expressed with the Fc region of human IgG1 at the C-terminus.