Rat Recombinant TrkA/NTRK1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80404-R02H-100
103584-884EA 300.98 USD
103584-884
Rat Recombinant TrkA/NTRK1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat NTRK1 (P35739-Isoform TrkA-II)(Met1-Pro418) was expressed with the Fc region of human IgG1 at the C-terminus.