Rat Recombinant JAM-2/JAM-B Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80376-R02H-100
103584-874EA 300.98 USD
103584-874
Rat Recombinant JAM-2/JAM-B Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat JAM2 (Q3MHC0) (Met1-Asn236) was expressed, fused with the Fc region of human IgG1 at the C-terminus.