Rat Recombinant c-MPL/CD110/TPOR Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
80346-R08B-100
103584-842EA 300.98 USD
103584-842
Rat Recombinant c-MPL/CD110/TPOR Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of rat MPL (Met1-Ala500) was expressed with a polyhistide tag at the C-terminus.