Rat Recombinant CD14 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80308-R02H-200 80308-R02H-100
103666-622EA 320.35 USD
103666-622 103666-620
Rat Recombinant CD14 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat CD14 (Q63691) (Met1-Tyr341) was expressed, fused with the Fc region of human IgG1 at the C-terminus.