Rat Recombinant Cadherin-6/CDH6 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80277-R08H-200
103666-572EA 401.98 USD
103666-572
Rat Recombinant Cadherin-6/CDH6 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat CDH6 (EDL82944.1) (Met1-Ala615) was expressed with a polyhistidine tag at the C-terminus.